Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WFDC9
Entrez Gene ID
102190147
Gene Name
WAP four-disulfide core domain 9
Gene Aliases
-
Chromosome Location
Chr.13: 73947776 - 73948718 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005688717.1 XP_005688774.1 1 - 2 86 64

View Full Product Details