Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ODF3L1
Entrez Gene ID
102189247
Gene Name
outer dense fiber of sperm tails 3 like 1
Gene Aliases
-
Chromosome Location
Chr.21: 32611097 - 32615184 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005695118.2 XP_005695175.2 3 - 4 346 74

View Full Product Details