Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR82
Entrez Gene ID
102184565
Gene Name
WD repeat domain 82
Gene Aliases
-
Chromosome Location
Chr.22: 48718134 - 48735556 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018066939.1 XP_017922428.1 8 - 9 1178 63
GenBank mRNA JO359473.2 - - 2119 63
JO579201.1 - 1 - 2 2950 63

View Full Product Details