Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DCST2
Entrez Gene ID
102169237
Gene Name
DC-STAMP domain containing 2
Gene Aliases
-
Chromosome Location
Chr.3: 104557105 - 104570818 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018046213.1 XP_017901702.1 4 - 5 740 76

View Full Product Details