Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CALM2
- Entrez Gene ID
-
102172164
- Gene Name
- calmodulin 2
- Gene Aliases
-
-
- Chromosome Location
-
Chr.11: 29156106 - 29169683 on Build ARS1
- Gene Symbol
- LRRC8A
- Entrez Gene ID
-
102179957
- Gene Name
- leucine rich repeat containing 8 VRAC subunit A
- Gene Aliases
-
-
- Chromosome Location
-
Chr.11: 98867723 - 98895376 on Build ARS1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
JO367595.2 |
- |
- |
74 |
82 |
View Full Product Details