Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPINK8
Entrez Gene ID
102191657
Gene Name
serine peptidase inhibitor, Kazal type 8 (putative)
Gene Aliases
-
Chromosome Location
Chr.22: 51600129 - 51608402 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013973489.2 XP_013828943.1 3 - 4 262 72

View Full Product Details