Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SGF29
Entrez Gene ID
102188556
Gene Name
SAGA complex associated factor 29
Gene Aliases
CCDC101
Chromosome Location
Chr.25: 26000052 - 26032434 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018040164.1 XP_017895653.1 4 - 5 423 104
XM_018040165.1 XP_017895654.1 5 - 6 463 104

View Full Product Details