Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCTD2
Entrez Gene ID
102175161
Gene Name
potassium channel tetramerization domain containing 2
Gene Aliases
-
Chromosome Location
Chr.19: 55603032 - 55617060 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018063838.1 XP_017919327.1 5 - 6 795 82
GenBank mRNA KA317848.1 - 1 - 2 2629 82

View Full Product Details