Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GPAT3
Entrez Gene ID
102180745
Gene Name
glycerol-3-phosphate acyltransferase 3
Gene Aliases
AGPAT9
Chromosome Location
Chr.6: 98745864 - 98810284 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018049681.1 XP_017905170.1 4 - 5 1122 84
GenBank mRNA JO380617.2 - 3 - 4 631 84

View Full Product Details