Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fsn
Entrez Gene ID
36460
Gene Name
F-box synaptic protein
Gene Aliases
BEST:GH20696, CG4643, DFsn, Dmel\CG4643, Dmel_CG4643, FBXO2, Wals, dFsn, fsn
Chromosome Location
Chr.2R: 13220939 - 13222459 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0043010
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_137005.2 NP_610849.1 2 - 3 486 66
NM_165973.2 NP_725266.1 1 - 2 479 66
GenBank mRNA AY118594.1 - 1 - 2 459 66

View Full Product Details