Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
scaf6
Entrez Gene ID
3346235
Gene Name
SR-related CTD associated factor 6
Gene Aliases
BP1040, BP1047, CG14058, CG32168, CG33522, CG6615, CG6626, Dmel\CG33522, Dmel_CG33522
Chromosome Location
Chr.3L: 17036859 - 17042574 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0261872
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014589.4 NP_001014589.1 10 - 11 2412 111
NM_001038939.2 NP_001034028.2 9 - 10 2339 111
GenBank mRNA BT031129.1 - 10 - 11 2378 111

View Full Product Details