Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tra2
Entrez Gene ID
36619
Gene Name
transformer 2
Gene Aliases
CG10128, Dmel\CG10128, Dmel_CG10128, TRA 2, TRA-2, TRA2, Tra-2, Tra2, tra-2
Chromosome Location
Chr.2R: 14602004 - 14604337 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0003742
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_057415.3 NP_476763.1 3 - 4 433 52
NM_057416.3 NP_476764.1 3 - 4 433 52
NM_057417.3 NP_476765.1 4 - 5 625 52
NM_057418.2 NP_476766.1 1 - 2 517 52
NM_134280.1 NP_599107.1 2 - 3 285 52
NM_206112.2 NP_995834.1 3 - 4 448 52
NM_206113.2 NP_995835.1 3 - 4 438 52
GenBank mRNA M23633.1 - 3 - 4 325 52
M76381.1 - 3 - 4 453 52

View Full Product Details