Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ssp3
Entrez Gene ID
35149
Gene Name
short spindle 3
Gene Aliases
CG10666, CG18397, Dmel\CG18397, Dmel_CG18397, Ssp3
Chromosome Location
Chr.2L: 18916347 - 18943960 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0032723
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001299140.1 NP_001286069.1 10 - 11 5577 68
NM_136073.3 NP_609917.2 10 - 11 5583 68
GenBank mRNA BT021951.1 - 10 - 11 5567 68

View Full Product Details