Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
meso18E
Entrez Gene ID
53448
Gene Name
CG14233-PA
Gene Aliases
CG14233, Dmel\CG14233, Dmel_CG14233, Dmeso18E, Meso18E, Mmeso18E, anon-18DEd, eso18E, ld26007
Chromosome Location
Chr.X: 19706418 - 19713045 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0040089
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169328.2 NP_001162799.1 4 - 5 2053 57
NM_001298531.1 NP_001285460.1 5 - 6 1545 57
NM_001298532.1 NP_001285461.1 4 - 5 1576 57
NM_144358.3 NP_652615.1 4 - 5 1478 57
GenBank mRNA AJ271814.1 - 4 - 5 2006 57
AY058570.1 - 4 - 5 1431 57

View Full Product Details