Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tsp42Ea
Entrez Gene ID
59178
Gene Name
Tetraspanin 42Ea
Gene Aliases
BcDNA:GH05668, BcDNA:GM15292, CG18742, CG18817, CG3364, Dm.Tsp42Ea, Dmel\CG18817, Dmel_CG18817, GH05668, Q9U3V4
Chromosome Location
Chr.2R: 6992676 - 7004259 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0029508
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080273.3 NP_525012.2 3 - 4 410 65
NM_144389.2 NP_652646.1 3 - 4 405 65
NM_165502.2 NP_724524.1 3 - 4 399 65
GenBank mRNA AF274014.1 - 3 - 4 397 65
AY069049.1 - 3 - 4 397 65

View Full Product Details