Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tfb1
Entrez Gene ID
36598
Gene Name
Transcription factor B1
Gene Aliases
CG8151, Dmel\CG8151, Dmel_CG8151, Dmp62, TFB1, TfBl, l(2)06949, l(2)06950, p62
Chromosome Location
Chr.2R: 14363048 - 14365473 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0033929
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001259392.2 NP_001246321.1 2 - 3 288 63
NM_137113.4 NP_610957.1 2 - 3 295 63
NM_166048.1 NP_725382.1 1 - 2 415 63
NM_166049.1 NP_725383.1 2 - 3 292 63
GenBank mRNA AY052097.1 - 1 - 2 273 63

View Full Product Details