Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bsg
Entrez Gene ID
318841
Gene Name
Basigin
Gene Aliases
135/10, BSG, BcDNA:GH21853, CG31605, Dmel\CG31605, Dmel_CG31605, NP6293, bsg, gel, l(2)06243, l(2)SH1217, l(2)SH2 1217, l(2)k06338, l(2)k09030, l(2)k13638, l(2)k14308, ms(2)08318
Chromosome Location
Chr.2L: 8083420 - 8116531 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0261822
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273295.1 NP_001260224.1 2 - 3 709 115
NM_164782.2 NP_723338.1 2 - 3 567 115
NM_164783.2 NP_723339.1 2 - 3 452 115
NM_164784.2 NP_723340.1 2 - 3 776 115
NM_164785.2 NP_723341.1 2 - 3 661 115
NM_164786.2 NP_723342.1 2 - 3 500 115
NM_164787.2 NP_723343.1 3 - 4 805 115
NM_164788.2 NP_723344.1 2 - 3 709 115
NM_164789.3 NP_723345.1 1 - 2 641 115
GenBank mRNA BT005198.1 - 2 - 3 414 115
BT006333.1 - 2 - 3 616 115

View Full Product Details