Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Myo31DF
Entrez Gene ID
34445
Gene Name
Myosin 31DF
Gene Aliases
CG7438, Dm IA, DmIA, Dmel\CG7438, Dmel_CG7438, DroMIA, IA, MYOIA, Mhc1, Myo-IA, Myo1A, Myo1D, MyoIA, MyoID, myo31DF, myoID, souther
Chromosome Location
Chr.2L: 10491814 - 10506779 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0086347
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001201855.2 NP_001188784.1 5 - 6 1168 75
NM_078814.4 NP_523538.1 5 - 6 1027 75
NM_164933.3 NP_723596.1 4 - 5 1604 75
GenBank mRNA AY075567.1 - 4 - 5 1583 75
U07595.1 - 5 - 6 1041 75

View Full Product Details