Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Oatp74D
Entrez Gene ID
39954
Gene Name
Organic anion transporting polypeptide 74D
Gene Aliases
CG7571, Dmel\CG7571, Dmel_CG7571, EcI, oatp 74D
Chromosome Location
Chr.3L: 17487118 - 17511693 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0036732
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169989.2 NP_001163460.1 4 - 5 2061 58
NM_001300166.1 NP_001287095.1 4 - 5 2004 58
NM_001300167.1 NP_001287096.1 3 - 4 2409 58
NM_140732.3 NP_648989.1 4 - 5 2050 58
GenBank mRNA BT001436.1 - 4 - 5 2029 58

View Full Product Details