Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CenG1A
Entrez Gene ID
34803
Gene Name
Centaurin gamma 1A
Gene Aliases
AGAP, BG:DS08220.1, CG16866, CG16867, CG31811, CG7982, CenG1a, Dmel\CG31811, Dmel_CG31811, cenG, cenG1A, cenG1a, ceng1A
Chromosome Location
Chr.2L: 13835564 - 13898712 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0028509
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078838.3 NP_523562.2 6 - 7 2660 69
NM_165067.2 NP_723849.1 6 - 7 2779 69
NM_165068.2 NP_723850.1 5 - 6 1851 69
GenBank mRNA AY122158.1 - 6 - 7 2642 69
BT053695.1 - 5 - 6 1829 69
BT133428.1 - 6 - 7 2776 69

View Full Product Details