Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- dac
- Entrez Gene ID
-
34982
- Gene Name
- dachshund
- Gene Aliases
-
BG:DS02780.3, CG4952, DAC, Dac, Dac2-3, Dach, Dm-DAC, Dmel\CG4952, Dmel_CG4952, dach, l(2)36Ae, l(2)rK364, spn
- Chromosome Location
-
Chr.2L: 16466512 - 16485998 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0005677
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_165162.3 |
NP_723971.1 |
3 - 4 |
1589 |
61 |
View Full Product Details