Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- rho-5
- Entrez Gene ID
-
2768944
- Gene Name
- rhomboid-5
- Gene Aliases
-
CG33304, CG5364, CT17046, Dmel\CG33304, Dmel_CG33304, Rho-related[31D10], iRhom, iRhom1
- Chromosome Location
-
Chr.2L: 10368025 - 10373900 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0041723
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_205957.3 |
NP_995679.1 |
5 - 6 |
3968 |
67 |
View Full Product Details