Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
spir
Entrez Gene ID
45931
Gene Name
spire
Gene Aliases
38C.34, 38C.37, CG10076, CG18621, Dmel\CG10076, Dmel_CG10076, NP2788, Spir, Spire, l(2)08327, p150-Spir, p150Spir
Chromosome Location
Chr.2L: 20311219 - 20348386 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0003475
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169549.1 NP_001163020.1 12 - 13 2498 63
NM_001259172.2 NP_001246101.1 13 - 14 2582 63
NM_001299171.1 NP_001286100.1 13 - 14 3305 63
NM_080115.3 NP_524854.2 12 - 13 2501 63
NM_165323.3 NP_724254.1 13 - 14 2588 63
NM_165325.3 NP_724256.2 5 - 6 1144 63
GenBank mRNA AJ238876.1 - 13 - 14 2235 63
BT044175.1 - 5 - 6 1119 63
BT122081.1 - 13 - 14 2981 63

View Full Product Details