Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- hubl
- Entrez Gene ID
-
246567
- Gene Name
- hug-bell
- Gene Aliases
-
CG2149, CG30364, Dmel\CG30364, Dmel_CG30364, anon-WO0140519.168
- Chromosome Location
-
Chr.2R: 8359466 - 8362156 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0050364
- Gene Symbol
- spaw
- Entrez Gene ID
-
246568
- Gene Name
- spacewatch
- Gene Aliases
-
BcDNA:AT04457, CG2149, CG30365, Dmel\CG30365, Dmel_CG30365
- Chromosome Location
-
Chr.2R: 8359466 - 8362156 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0050365
View Full Product Details