Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Or13a
- Entrez Gene ID
-
32562
- Gene Name
- Odorant receptor 13a
- Gene Aliases
-
13F.1, 13a, CG12697, DOR38, Dmel\CG12697, Dmel_CG12697, dOr13a, or13a
- Chromosome Location
-
Chr.X: 15974425 - 15976706 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0030715
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078635.3 |
NP_523359.2 |
3 - 4 |
876 |
107 |
View Full Product Details