Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tsp39D
Entrez Gene ID
35405
Gene Name
Tetraspanin 39D
Gene Aliases
CG8666, Dm.Tsp39D, Dmel\CG8666, Dmel_CG8666
Chromosome Location
Chr.2L: 21344607 - 21358271 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0032943
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078888.4 NP_523612.3 2 - 3 248 91
GenBank mRNA BT010223.1 - 2 - 3 228 91

View Full Product Details