Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- GstT1
- Entrez Gene ID
-
35995
- Gene Name
- Glutathione S transferase T1
- Gene Aliases
-
BcDNA:RE63575, CG12930, CG12930B, CG30000, DmGSTT1, Dmel\CG30000, Dmel_CG30000, GSTT1, gstt1
- Chromosome Location
-
Chr.2R: 9605988 - 9607184 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0050000
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_136665.5 |
NP_610509.2 |
2 - 3 |
323 |
63 |
GenBank mRNA |
AY070580.1 |
- |
2 - 3 |
321 |
63 |
View Full Product Details