Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- RpI1
- Entrez Gene ID
-
36617
- Gene Name
- RNA polymerase I subunit
- Gene Aliases
-
153129_at, CG10122, DmRPA1, Dmel\CG10122, Dmel_CG10122, RPA1
- Chromosome Location
-
Chr.2R: 14595611 - 14601216 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0019938
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079019.3 |
NP_523743.1 |
2 - 3 |
810 |
75 |
View Full Product Details