Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
enok
Entrez Gene ID
37859
Gene Name
enoki mushroom
Gene Aliases
CG11290, Cg11290, Dmel\CG11290, Dmel_CG11290, Enok, KAT6, S1, anon-60Ba, anon-WO0200864.1, anon-WO0200864.2, dmHAG406, rot
Chromosome Location
Chr.2R: 24099210 - 24107032 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0034975
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001299894.1 NP_001286823.1 3 - 4 1386 101
NM_079114.2 NP_523838.1 3 - 4 1117 101

View Full Product Details