Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lcp9
Entrez Gene ID
37963
Gene Name
Larval cuticle protein 9
Gene Aliases
CG16914, DmelLcp9, Dmel\CG16914, Dmel_CG16914, LCP9, LCPX, lcp9
Chromosome Location
Chr.2R: 24779979 - 24780428 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0025578
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_079130.2 NP_523854.1 2 - 2 293 100
GenBank mRNA BT100156.4 - 1 - 1 234 100
KX531500.1 - 1 - 1 234 100

View Full Product Details