Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
bab2
Entrez Gene ID
44254
Gene Name
bric a brac 2
Gene Aliases
BAB, BAB2, BTB-II, Bab2, BtbII, CG13911, CG9102, Dmel\CG9102, Dmel_CG9102, bab, bab-II
Chromosome Location
Chr.3L: 1140429 - 1177276 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0025525
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001274313.1 NP_001261242.1 2 - 3 2145 61
NM_079155.3 NP_523879.2 2 - 3 2145 61

View Full Product Details