Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Arp3
Entrez Gene ID
38898
Gene Name
Actin-related protein 3
Gene Aliases
ARP3, AcTr66B, Actr66B, Arp66B, CG7558, Dmel\CG7558, Dmel_CG7558, anon-WO0172774.138, arp3, arp66B, arp66b, schwachling
Chromosome Location
Chr.3L: 8113407 - 8115902 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0262716
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001274630.1 NP_001261559.1 5 - 6 1345 65
NM_079244.3 NP_523968.1 5 - 6 1345 65
GenBank mRNA AY051859.1 - 5 - 6 1315 65

View Full Product Details