Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- rt
- Entrez Gene ID
-
39297
- Gene Name
- rotated abdomen
- Gene Aliases
-
CG6097, DPOMT1, Dmel\CG6097, Dmel_CG6097, POMT-1, POMT1, Rot_abd, Rt, dPOMT1, dPomt1, fs(3)neo1, pomt, roa
- Chromosome Location
-
Chr.3L: 11584122 - 11587770 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0003292
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079301.4 |
NP_524025.2 |
1 - 2 |
1033 |
74 |
View Full Product Details