Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Nrx-IV
- Entrez Gene ID
-
39387
- Gene Name
- Neurexin IV
- Gene Aliases
-
CG682, CG6827, CNTNAP2, CT21123, Dmel\CG6827, Dmel_CG6827, Dnrx, NRX, NRX IV, Nrx, Nrx IV, NrxIV, Nx IV, l(3)68Ff, l(3)rsg23, nrx, nrx IV, nrx-IV, nrxIV, rsg23
- Chromosome Location
-
Chr.3L: 12146519 - 12156990 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0013997
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079310.3 |
NP_524034.2 |
3 - 4 |
753 |
68 |
View Full Product Details