Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
G9a
Entrez Gene ID
30971
Gene Name
CG2995-PA
Gene Aliases
CG2995, Dmel\CG2995, Dmel_CG2995, EG:BACR37P7.2, EHMT, cab65850 G9a Dm, dG9a
Chromosome Location
Chr.X: 245978 - 254650 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0040372
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001272159.1 NP_001259088.1 5 - 6 4331 65
NM_130478.3 NP_569834.1 5 - 6 4331 65
GenBank mRNA AY061125.1 - 5 - 6 4302 65

View Full Product Details