Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tpst
Entrez Gene ID
32312
Gene Name
Tyrosylprotein sulfotransferase
Gene Aliases
CG1573, CG15758, CG15759, CG32629, CG32629/CG32632, CG32632, DmTPST, Dmel\CG32632, Dmel_CG32632, TANGO13, TPST, Tango13
Chromosome Location
Chr.X: 13563823 - 13593224 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0086674
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103503.2 NP_001096973.1 4 - 5 1980 65
NM_001272589.1 NP_001259518.1 3 - 4 1332 65
NM_001272590.1 NP_001259519.1 4 - 5 1980 65
NM_001272591.1 NP_001259520.1 4 - 5 1980 65
NM_001272592.1 NP_001259521.1 4 - 5 2174 65
NM_167374.3 NP_727717.1 3 - 4 1332 65
GenBank mRNA AY119073.1 - 2 - 3 370 65
AY124548.1 - 2 - 3 370 65
BT021950.1 - 4 - 5 1957 65

Gene Symbol
CG46311
Entrez Gene ID
30522400
Gene Name
CG46311-PA
Gene Aliases
Dmel\CG46311, Dmel_CG46311, Tango13-5'utr
Chromosome Location
Chr.X: 13563823 - 13590570 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0284227
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001347789.1 NP_001334677.1 4 - 5 1980 65

View Full Product Details