Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
acj6
Entrez Gene ID
47080
Gene Name
abnormal chemosensory jump 6
Gene Aliases
ACJ6, Acj6, CG9151, Dmel\CG9151, Dmel_CG9151, I-POU, Ipou, tI-POU
Chromosome Location
Chr.X: 15350239 - 15379064 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0000028
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014743.3 NP_001014743.1 6 - 7 1486 74
NM_001014744.2 NP_001014744.1 5 - 6 1408 74
NM_001144726.2 NP_001138198.1 4 - 5 1396 74
NM_001258753.2 NP_001245682.1 6 - 7 1465 74
NM_001258754.2 NP_001245683.1 4 - 5 1375 74
NM_001258755.2 NP_001245684.1 4 - 5 1396 74
NM_001258756.2 NP_001245685.1 4 - 5 1375 74
NM_001258757.2 NP_001245686.1 5 - 6 1408 74
NM_001258758.2 NP_001245687.1 5 - 6 1432 74
NM_001258759.2 NP_001245688.1 5 - 6 1453 74
NM_001258760.2 NP_001245689.1 5 - 6 1432 74
NM_080137.3 NP_524876.1 6 - 7 1486 74
NM_167438.2 NP_727845.1 5 - 6 1429 74
GenBank mRNA AF086816.1 - 6 - 7 876 74

View Full Product Details