Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cerv
Entrez Gene ID
32492
Gene Name
cervantes
Gene Aliases
CG15645, Cerv, Dmel\CG15645, Dmel_CG15645
Chromosome Location
Chr.X: 15576986 - 15578171 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0030657
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001298349.1 NP_001285278.1 2 - 2 981 64
NM_132815.3 NP_573043.1 2 - 2 808 64
NM_167453.3 NP_727868.1 2 - 2 792 64
GenBank mRNA BT011022.1 - 2 - 2 794 64

View Full Product Details