Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Vps26
- Entrez Gene ID
-
31144
- Gene Name
- Vacuolar protein sorting 26
- Gene Aliases
-
131F2S, CG14804, DmVps26, Dmel\CG14804, Dmel_CG14804, Dvps26, EG:63B12.8, ESTS:131F2S, VPS26, anon-sts13, dVps26, dvps26, vps26
- Chromosome Location
-
Chr.X: 1873853 - 1876474 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0014411
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_130596.3 |
NP_569952.2 |
1 - 2 |
433 |
82 |
GenBank mRNA |
BT023502.1 |
- |
1 - 2 |
423 |
82 |
View Full Product Details