Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Fmo-1
- Entrez Gene ID
-
37814
- Gene Name
- Flavin-containing monooxygenase 1
- Gene Aliases
-
CG3006, DmFMO-1, Dmel\CG3006, Dmel_CG3006, Dmfmo1, FMO-1
- Chromosome Location
-
Chr.2R: 23962956 - 23964641 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0034943
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_138015.4 |
NP_611859.1 |
3 - 4 |
643 |
67 |
GenBank mRNA |
AY051603.1 |
- |
3 - 4 |
624 |
67 |
View Full Product Details