Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- snama
- Entrez Gene ID
-
37857
- Gene Name
- something that sticks like glue
- Gene Aliases
-
BCDNA:LD21643, BcDNA:LD21643, CG3231, Dmel\CG3231, Dmel_CG3231, FBgn0086129, SNAMA, Snama, l(2)rQ313, mnm
- Chromosome Location
-
Chr.2R: 24086604 - 24092014 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0086129
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_138040.4 |
NP_611884.1 |
7 - 8 |
1752 |
61 |
GenBank mRNA |
AF132177.1 |
- |
7 - 8 |
1724 |
61 |
View Full Product Details