Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ens
Entrez Gene ID
38491
Gene Name
ensconsin
Gene Aliases
CG14998, Dm-308, Dmel\CG14998, Dmel_CG14998, Ens, ensc, swoosh
Chromosome Location
Chr.3L: 4122907 - 4134550 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0264693
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104028.2 NP_001097498.1 4 - 5 514 78
NM_001274470.1 NP_001261399.1 3 - 4 397 78
NM_001274471.1 NP_001261400.1 3 - 4 397 78
NM_001274472.1 NP_001261401.1 3 - 4 397 78
NM_001274473.1 NP_001261402.1 3 - 4 397 78
NM_001274474.1 NP_001261403.1 3 - 4 397 78
NM_001300004.1 NP_001286933.1 3 - 4 406 78
NM_139604.2 NP_647861.2 2 - 3 352 78
NM_168061.2 NP_728938.2 3 - 4 406 78
NM_168062.3 NP_728939.2 3 - 4 406 78
NM_168063.2 NP_728940.2 3 - 4 406 78
NM_168064.2 NP_728941.2 4 - 5 514 78
GenBank mRNA BT010301.1 - 4 - 5 472 78
BT125906.1 - 3 - 4 392 78
BT126155.1 - 3 - 4 387 78

View Full Product Details