Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Elo68alpha
Entrez Gene ID
317841
Gene Name
Elongase 68alpha
Gene Aliases
CG32072, CG6261, Dmel\CG32072, Dmel_CG32072, elo68alpha
Chromosome Location
Chr.3L: 11101875 - 11103030 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0052072
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_168440.3 NP_729666.2 1 - 2 182 73
GenBank mRNA AJ871925.1 - 1 - 2 183 73
BT022934.1 - 1 - 2 175 73
BT023054.1 - 1 - 2 182 73

View Full Product Details