Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Set2
Entrez Gene ID
32301
Gene Name
SET domain containing 2
Gene Aliases
CG1716, Dmel\CG1716, Dmel_CG1716, Hypb, SET2, dHypb, dSet2, set2
Chromosome Location
Chr.X: 13413242 - 13421024 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0030486
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169269.2 NP_001162740.1 2 - 3 4066 84
NM_132660.4 NP_572888.2 1 - 2 4277 84
GenBank mRNA BT025042.1 - 2 - 3 4051 84

View Full Product Details