Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Eo
Entrez Gene ID
32423
Gene Name
Ecdysone oxidase
Gene Aliases
CG9504, DmEO, Dmel\CG9504, Dmel_CG9504, EO, EObeta1
Chromosome Location
Chr.X: 14916756 - 14918800 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0030597
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_132758.2 NP_572986.1 1 - 1 1993 81
GenBank mRNA BT100214.1 - 1 - 1 1937 81

View Full Product Details