Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tud
Entrez Gene ID
37417
Gene Name
tudor
Gene Aliases
CG9450, Dmel\CG9450, Dmel_CG9450, R5_2R:17070527..17070890, TUD, Tud, Tudor, tu
Chromosome Location
Chr.2R: 21183322 - 21192913 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0003891
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001299736.1 NP_001286665.1 8 - 9 4541 93
NM_057425.3 NP_476773.1 8 - 9 4550 93

View Full Product Details