Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gr2a
Entrez Gene ID
31093
Gene Name
Gustatory receptor 2a
Gene Aliases
CG18531, Dmel\CG18531, Dmel_CG18531, EG:BACN32G11.1, GR1F.1, GR2B1, GR2a, Gr1F1, Gr2B1
Chromosome Location
Chr.X: 1452430 - 1454177 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0265139
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169162.2 NP_001162633.1 3 - 4 1406 79
NM_080299.4 NP_525038.3 4 - 5 1352 79
GenBank mRNA BT122077.2 - 3 - 4 1150 79

View Full Product Details