Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rhi
Entrez Gene ID
44879
Gene Name
rhino
Gene Aliases
CG10683, Dmel\CG10683, Dmel_CG10683, HP1, HP1D, HP1d, RHI, Rhi, Rhino, hp1d, rno
Chromosome Location
Chr.2R: 17628491 - 17633516 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0004400
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080533.4 NP_536794.1 2 - 3 1317 101
GenBank mRNA AF411862.1 - 2 - 3 1301 101
AY094869.1 - 2 - 3 1314 101

View Full Product Details