Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Obp18a
- Entrez Gene ID
-
32897
- Gene Name
- Odorant-binding protein 18a
- Gene Aliases
-
18a, BcDNA:RH25665, CG15883, DmelObp18a, Dmel\CG15883, Dmel_CG15883, obp18a
- Chromosome Location
-
Chr.X: 19135081 - 19135868 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0030985
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_133122.2 |
NP_573350.1 |
1 - 2 |
76 |
85 |
GenBank mRNA |
KX531605.1 |
- |
1 - 2 |
51 |
85 |
View Full Product Details