Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- cutlet
- Entrez Gene ID
-
44637
- Gene Name
- CG33122-PA
- Gene Aliases
-
CG10030, CG33122, Dmel\CG33122, Dmel_CG33122, ctl
- Chromosome Location
-
Chr.2L: 3707230 - 3711748 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0015376
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_175955.3 |
NP_787969.1 |
1 - 2 |
1007 |
67 |
View Full Product Details